Skip to Content
Merck
All Photos(1)

Key Documents

W352306

Sigma-Aldrich

Pyrrolidine

≥99%

Synonym(s):

Tetrahydropyrrole, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
FEMA Number:
3523
Beilstein:
102395
EC Number:
Council of Europe no.:
10491
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.064
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens, no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Kosher

vapor density

2.45 (vs air)

vapor pressure

128 mmHg ( 39 °C)
49 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

653 °F

expl. lim.

10.6 %

refractive index

n20/D 1.443 (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens, no known allergens

Organoleptic

fishy

SMILES string

C1CCNC1

InChI

1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2

InChI key

RWRDLPDLKQPQOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor A Kirilyuk et al.
The Journal of organic chemistry, 77(18), 8016-8027 (2012-08-25)
The nitroxides of 7-azadispiro[5.1.5.2]pentadecane and 7-azadispiro[5.1.5.2]pentadeca-14-ene series have been prepared, including thiol-specific methane thiosulfonate spin label for site-directed spin labeling. The effect of spirocyclohexane moieties on chemical and spectral properties has been studied. The obtained temperature dependencies of electron spin
Boyu Zhang et al.
Angewandte Chemie (International ed. in English), 51(52), 13159-13162 (2012-11-20)
Drop it! A highly enantioselective catalytic cascade reaction of α-ketoacids and aldehydes is achieved using the title catalyst and water as the solvent. Fluorescence imaging shows that the catalyst is mainly distributed on the surface of emulsion droplets. Optically active
Yukari Kondo et al.
The Journal of organic chemistry, 77(18), 7988-7999 (2012-08-18)
We report an enantioselective synthesis of the polyhydroxylated pyrrolidine alkaloid (+)-DMDP. The key steps in the synthesis were guanidinium ylide mediated asymmetric aziridination, stereospecific ring opening of trans-3-vinylaziridine-2-carboxylate with an oxygen nucleophile, iodine-mediated 5-endo-trig amino cyclization, and Prévost displacement. In
Matias Rey-Carrizo et al.
Journal of medicinal chemistry, 56(22), 9265-9274 (2013-11-19)
We have synthesized and characterized a series of compounds containing the 3-azatetracyclo[5.2.1.1(5,8).0(1,5)]undecane scaffold designed as analogues of amantadine, an inhibitor of the M2 proton channel of influenza A virus. Inhibition of the wild-type (WT) M2 channel and the amantadine-resistant A/M2-S31N
Marius Retegan et al.
Physical chemistry chemical physics : PCCP, 15(1), 223-234 (2012-11-20)
The electronic and magnetic properties of polynuclear complexes, in particular the magnetic anisotropy (zero field splitting, ZFS), the leading term of the spin Hamiltonian (SH), are commonly analyzed in a global manner and no attempt is usually made to understand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service