Skip to Content
Merck
All Photos(1)

Key Documents

W305804

Sigma-Aldrich

DL-Tetrahydrofurfuryl propionate

≥97%

Synonym(s):

N-Propionic acid tetrahydrofurfuryl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
FEMA Number:
3058
EC Number:
Council of Europe no.:
2096
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.049
Pricing and availability is not currently available.

biological source

synthetic

grade

Kosher

Assay

≥97%

refractive index

n20/D 1.438 (lit.)

bp

207 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

caramel; medicinal; fruity; earthy

SMILES string

CCC(=O)OCC1CCCO1

InChI

1S/C8H14O3/c1-2-8(9)11-6-7-4-3-5-10-7/h7H,2-6H2,1H3

InChI key

FMKCDSXOYLTWBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tropical fruits such as mango, berry nuances, banana, rum and malt flavors.

Features and Benefits

Odor: @ 10.0 %. Weak, musty, sweetly ethereal, tropical rummy, solvent winey and fermented, with an overripe fruity banana nuance.
Taste: @ 50 to 100 ppm. Musty, solventy, fruity, slightly fermented, rummy, malt-like with an overripe fruit nuance.
Fruity, earthy, phenolic, medicinal

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 104-104 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service