Skip to Content
Merck
All Photos(1)

Documents

W220507

Sigma-Aldrich

Butyl (S)-(−)-lactate

≥97%

Synonym(s):

(−)-Butyl L-lactate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2(CH2)3CH3
CAS Number:
Molecular Weight:
146.18
FEMA Number:
2205
Beilstein:
1721597
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.434

biological source

synthetic

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

vapor density

4 (20 °C, vs air)

Assay

≥97%

optical activity

[α]20/D −12°, neat

autoignition temp.

720 °F

bp

185-187 °C (lit.)

mp

−28 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

creamy; sweet

SMILES string

CCCCOC(=O)[C@H](C)O

InChI

1S/C7H14O3/c1-3-4-5-10-7(9)6(2)8/h6,8H,3-5H2,1-2H3/t6-/m0/s1

InChI key

MRABAEUHTLLEML-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service