Skip to Content
Merck
All Photos(1)

Key Documents

W203009

Sigma-Aldrich

Allyl 2-furoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
FEMA Number:
2030
EC Number:
Council of Europe no.:
360
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.004
NACRES:
NA.21
To inquire about this product W203009, please contact your local Merck office or dealer. Contact Technical Service

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥98%

impurities

≤890 ppm Toluene (residual solvent)

refractive index

n20/D 1.495 (lit.)

bp

206-209 °C (lit.)

density

1.181 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; fruity

SMILES string

C=CCOC(=O)c1ccco1

InChI

1S/C8H8O3/c1-2-5-11-8(9)7-4-3-6-10-7/h2-4,6H,1,5H2

InChI key

SJRQTHAMRUOPBJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service