Key Documents
TEM-CFT300CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 300 mesh, box of 50
Synonym(s):
CARBON THIN BAR SQUARE MESH - CU 300 MESH UL 50/BX
Select a Size
1 PKG
£158.00
£158.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
£158.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
£158.00
Please contact Customer Service for Availability
Request a Bulk Order
packaging
box of 50
size
300 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported thin bar square mesh grid. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)