Key Documents
TEM-CFLF135CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper London Finder LF135 Grids
box of 50
Synonym(s):
CARBON LONDON FINDER GRIDS - CU,LF135, ET 50/BX
Select a Size
1 PKG
£136.00
£136.00
Estimated to ship on07 May 2025Details
Request a Bulk Order
Select a Size
Change View
1 PKG
£136.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
£136.00
Estimated to ship on07 May 2025Details
Request a Bulk Order
packaging
box of 50
General description
A 3-4 nm ultrathin film of pure carbon supported London finder (LF135) grids. Copper grid. Carbon film show better electrical and heat conduction in the electron beam. London finder grids are locator grids with reference/index on the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)