TEM-CF300AUUL
Ultrathin Carbon Film Supported Gold Square Mesh
size 300 mesh, box of 50
Synonym(s):
CARBON SQUARE - MESH - AU 300 MESH UL 50/BX
All Photos(1)
About This Item
packaging
box of 50
size
300 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Gold grid, 300 mesh.