Key Documents
TEM-7435703
Sigma-Aldrich
Carbon Film Supported Copper Tomography Grid R14C2
pack of 25 ×
Synonym(s):
Carbon Film on Tomography Grid, 25pk
Select a Size
1 PKG
£313.00
£313.00
Estimated to ship on25 April 2025Details
Request a Bulk Order
Select a Size
Change View
1 PKG
£313.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
£313.00
Estimated to ship on25 April 2025Details
Request a Bulk Order
packaging
pack of 25 ×
General description
This 1.5mm square, 300 mesh grid is made from copper, supported by a carbon film, and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Choose from one of the most recent versions:
Certificates of Analysis (COA)