Key Documents
TEM-71150
Sigma-Aldrich
Grid Storage Box
capacity, 50 (wells), pkg of 1 ea
Synonym(s):
GRID STORAGE BOX 50-CAP CLEAR COVER R13A2
Select a Size
1 PKG
£3.80
£3.80
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
£3.80
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
£3.80
Please contact Customer Service for Availability
Request a Bulk Order
feature
capacity 50 (wells)
packaging
pkg of 1 ea
General description
Grid Storage Box, 50 CapacityStorage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Choose from one of the most recent versions:
Certificates of Analysis (COA)