Skip to Content
Merck
All Photos(1)

Key Documents

T75736

Sigma-Aldrich

2,2,6-Trimethylcyclohexanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H7(=O)
CAS Number:
Molecular Weight:
140.22
Beilstein:
1905593
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.447 (lit.)

bp

178-179 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC(C)(C)C1=O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service