Documents
SYNF464500
Synthware storage flask with standard taper side arm
capacity 500 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonym(s):
Synthware F464500, round flask with side arm
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F464500
capacity
500 mL
valve size
0-4 mm
General description
Supplied with high vacuum valve with PTFE O-ring at tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.