SLW1523/12
Pyrex® Media-Lab Bottles, wide mouth, with polypropylene cap and pouring ring, with printed trace code
Synonym(s):
Pyrex, SciLabware 1523/12
Select a Size
£397.00
Select a Size
About This Item
£397.00
Recommended Products
material
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
wide-mouth
Joint
Joints threaded neck (GLS80)
sterility
non-sterile
feature
graduations
with cap
with pouring ring
with printed trace code
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1523/12
H
252 mm
body diam.
136 mm
General description
- Bottles complete with polypropylene screwcap and drip-free pouring ring
- Manufactured from Pyrex borosilicate glass with proven temperature and chemical resistance qualities
- GLS 80 screwthread provides increased access compared to GL 45 bottles aiding filling and pouring
- Ideal for use with viscous liquids, pastes, powders, granules and larger solids
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- White enamel graduations and marking spot
- Autoclavable
- Screwcaps and pouring rings can withstand temperatures up to 140 deg. C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)