Key Documents
MT51340024
Mettler Toledo electrode adapter NS 14/15 to 12 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.79
manufacturer/tradename
Mettler-Toledo 51340024
General description
Adapter sleeve to NS 14.5 for sensors with 12–15 mm shaft diameter (material: PE)
Choose from one of the most recent versions:
Certificates of Analysis (COA)