Skip to Content
Merck
All Photos(1)

Key Documents

JKA13001

Sigma-Aldrich

mPEG6-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
£365.00

£365.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
£365.00

About This Item

Linear Formula:
CH3(CH2CH2O)5CH2CH2NH2
UNSPSC Code:
12162002
NACRES:
NA.23

£365.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

reaction suitability

reagent type: chemical modification reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Monodisperse PEG. The amine group reacts with a carboxyl group (-COOH) and other amine reactive chemical groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service