Key Documents
HAM201200
Hamilton® instrument syringe, for Boehringer Mannheim instruments, ES600
model, 1001, AccuDil (AD) end, volume 1 mL
Select a Size
1 EA
£117.00
£117.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
£117.00
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
£117.00
Please contact Customer Service for Availability
material
Borosilicate glass/PTFE
Gastight syringe
feature
autoclavable: no
model 1001, AccuDil (AD) end
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
200 psig pressure
volume
1 mL
Related Categories
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)