Skip to Content
Merck
All Photos(1)

Key Documents

H20105

Sigma-Aldrich

4-Hydroxybenzhydrazide

98%

Synonym(s):

4-Hydroxybenzoic hydrazide, 4-Hydroxybenzoylhydrazine, p-Hydroxybenzoic acid hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CONHNH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
473052
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

mp

264-266 °C (dec.) (lit.)

SMILES string

NNC(=O)c1ccc(O)cc1

InChI

1S/C7H8N2O2/c8-9-7(11)5-1-3-6(10)4-2-5/h1-4,10H,8H2,(H,9,11)

InChI key

ZMZGIVVRBMFZSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Israel Tilahun Desta et al.
The journal of physical chemistry. A, 124(2), 300-310 (2019-12-11)
Mechanical response of single crystals to light, temperature, and/or force-an emerging platform for the development of new organic actuating materials for soft robotics-has recently been quantitatively described by a general and robust mathematical model ( Chem. Rev . 2015

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service