Skip to Content
Merck
All Photos(1)

Documents

GF72398931

Zamak 410 - zinc/aluminum/copper/magnesium

foam, thickness 50 mm, Zn95%/Al 4%/Cu 1%/Mg%, size 150 × 150 mm, bulk density 0.33 g/cm3

Synonym(s):

ZN063802

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

Assay

≥95%

manufacturer/tradename

Goodfellow 723-989-31

size

150 × 150 mm

thickness

50 mm

bulk density

0.33 g/cm3

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service