GF62123813
Polypropylene
(PP), sheet, thickness 1.5 mm, size 300 × 300 mm, natural
Synonym(s):
PP303150
Select a Size
Vendor SKU: 5F-010-500MG
€628.30
Availability
Select a Size
About This Item
Vendor SKU: 5F-010-500MG
€628.30
Availability
Recommended Products
form
sheet
manufacturer/tradename
Goodfellow 621-238-13
size
300 × 300 mm
thickness
1.5 mm
color
natural
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service