Skip to Content
Merck
All Photos(1)

Key Documents

GF56129250

Titanium carbide

powder, mean particle size (fsss) 3 micron, weight 200 g, purity 98.5%

Synonym(s):

TI546010

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£698.00

£698.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
£698.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

£698.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98.5%

form

powder

manufacturer/tradename

Goodfellow 561-292-50

weight

200 g

mean particle size

3 μm (FSSR)

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service