Skip to Content
Merck
All Photos(2)

Documents

GF24883779

Gold

foil, thickness 0.015 mm, 8 mm diameter, purity 99.9%

Synonym(s):

AU000180

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 248-837-79

resistivity

2.05 μΩ-cm, 0°C

diam.

8 mm

thickness

0.015 mm

diameter

8 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service