Skip to Content
Merck
All Photos(1)

Key Documents

DWKBGE828PNC

Azlon Dropper Bottles with spouted closure and cap

round bottom, low-density polyethylene, capacity 150 mL

Synonym(s):

BGE828PNC

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
£15.90

£15.90


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
£15.90

About This Item

UNSPSC Code:
41121819
NACRES:
NB.43

£15.90


Please contact Customer Service for Availability

material

low-density polyethylene
round bottom

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

capacity

150 mL

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for as dropping bottle and excellent for dispensing gels

Features and Benefits

Pivotting nozzle closure

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service