DWK651483
Wheaton® Cryule Cryogenic Ampule
Synonym(s):
651483
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
preconditioned (pre-scored)
packaging
pkg of 144 ea
manufacturer/tradename
WHEATON® 651483
capacity
1.2 mL
diam. × H
11.8 mm × 58 mm
Looking for similar products? Visit Product Comparison Guide
Application
Biological Sample Preservation
Features and Benefits
- For biological sample preservation in glass
- Pre-scored to eliminate filing
- Allows for storage at low temperatures
- Can be used in the vapor phase of liquid nitrogen
- Special design allows for storage at low temperatures as well as rapid thawing
- Converted from Type I borosilicate glass tubing, ampules conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service