D140805
N-(4-Dimethylamino-3,5-dinitrophenyl)maleimide
97%
Synonym(s):
Tuppy’s maleimide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H10N4O6
CAS Number:
Molecular Weight:
306.23
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
97%
mp
177-181 °C (lit.)
SMILES string
CN(C)c1c(cc(cc1[N+]([O-])=O)N2C(=O)C=CC2=O)[N+]([O-])=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service