Skip to Content
Merck
All Photos(1)

Key Documents

CDS000383

Sigma-Aldrich

4-(Heptyloxy)benzaldehyde

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O2
CAS Number:
Molecular Weight:
220.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CCCCCCCOc1ccc(C=O)cc1

InChI

1S/C14H20O2/c1-2-3-4-5-6-11-16-14-9-7-13(12-15)8-10-14/h7-10,12H,2-6,11H2,1H3

InChI key

YBCKMIZXHKVONZ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalila Rocco et al.
Polymers, 12(2) (2020-02-09)
Five coordination polymers formed from combinations of copper(II) acetate and 4'-(4-alkyloxyphenyl)-3,2':6',3''-terpyridines with methoxy (1), n-butoxy (2), n-pentyloxy (3) and n-heptyloxy (4) substituents are reported. Reaction of 1 with Cu(OAc)2∙H2O leads to the 1D-polymer [Cu2(μ-OAc)4(1)]n in which {Cu2(-OAc)4} paddle-wheel units are
Naoko Ueno et al.
Langmuir : the ACS journal of surfaces and colloids, 33(22), 5393-5397 (2017-05-16)
We evaluated the speed profile of self-propelled underwater oil droplets comprising a hydrophobic aldehyde derivative in terms of their diameter and the surrounding surfactant concentration using a microfluidic device. We found that the speed of the oil droplets is dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service