Skip to Content
Merck
All Photos(1)

Key Documents

C51202

Sigma-Aldrich

5-Chloro-2-methylaniline

97%

Synonym(s):

2-Amino-4-chlorotoluene, 5-Chloro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
878504
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.584 (lit.)

bp

237 °C (lit.)

mp

20-22 °C (lit.)

density

1.177 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(Cl)cc1N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Massarelli et al.
European journal of medicinal chemistry, 44(9), 3658-3664 (2009-03-11)
A dataset comprising 55 chemicals with hepatocarcinogenic potency indices was collected from the Carcinogenic Potency Database with the aim of developing QSAR models enabling prediction of the above unwanted property for New Chemical Entities. The dataset was rationally split into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service