Skip to Content
Merck
All Photos(1)

Documents

C24507

Sigma-Aldrich

1-(4-Chlorobenzhydryl)piperazine

technical grade, 90%

Synonym(s):

(RS)-1-[(4-Chlorophenyl)phenylmethyl]piperazine, NSC 86164, Norchlorcyclizine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2
CAS Number:
Molecular Weight:
286.80
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

bp

178-180 °C/0.5 mmHg (lit.)

mp

65-70 °C (lit.)

SMILES string

Clc1ccc(cc1)C(N2CCNCC2)c3ccccc3

InChI

1S/C17H19ClN2/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-12-10-19-11-13-20/h1-9,17,19H,10-13H2

InChI key

UZKBSZSTDQSMDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service