Key Documents
BAH500250309
SP Bel-Art® Glove Box Bellows Type Gloves for 8 in. Glove Ports
latex, size 9 medium
Synonym(s):
glove box
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
latex
size
9 medium
feature
to hold Bellows and Gloves
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H50025-0309
General description
Made from natural latex with a removable hand, the sleeves fit 20cm (8") circular ports and 18 x 25cm (7 x 10") oval ports. Use with H50025-0320 Clamping Rings to hold gloves in place.
Features and Benefits
  • Forms a leak-proof unit with O-ring construction integral with the box
  • Bellows allows cooling air circulation
  • When the glove and sleeve are separated, the sleeve remains attached to the box
  • Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
  • Length of the extended sleeve and glove is approximately 76cm (30")
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)