Key Documents
BAH367180000
SP Bel-Art® Quaverette Micro-Blade Vibrating Spatula
stainless steel blade
Synonym(s):
spoon
Select a Size
1 EACH
£73.50
£73.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
£73.50
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
£73.50
Please contact Customer Service for Availability
material
stainless steel blade
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H36718-0000
General description
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Features and Benefits
Available with a plain stainless steel blade or with a brown FEP coating
Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)