ALD01906
Ethyl 6-oxo-4,5,6,7-tetrahydro-1H-indole-2-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
2-8°C
Application
This compound is a useful intermediate for the synthesis of biologically active compounds including pyrroloquinoline quinone analogs, which play a significant and productive role in a variety of mammalian processes, such as mitochondrial biogenesis and the attenuation of neurodegenerative diseases.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service