Skip to Content
Merck
All Photos(1)

Documents

ALD01692

Sigma-Aldrich

N-(4-(benzyloxy)-2-methylbutan-2-yl)-3-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

storage temp.

2-8°C

SMILES string

CC(C)(CCOCC1=CC=CC=C1)NC2=CC(C)=CC=C2

InChI

1S/C19H25NO/c1-16-8-7-11-18(14-16)20-19(2,3)12-13-21-15-17-9-5-4-6-10-17/h4-11,14,20H,12-13,15H2,1-3H3

InChI key

COYGPUDFQCNWDK-UHFFFAOYSA-N

Application

Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs. Additionally, this and over 80 other sterically hindered secondary amines in the Sigma-Aldrich portfolio provide structurally distinct screening compounds. Screen several through Aldrich Market Select. This hindered amine was accessed via a recent publication reported by Baran and coworkers using an innovative hydroamination method.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service