Skip to Content
Merck
All Photos(2)

Key Documents

ALD00562

Sigma-Aldrich

2,3-Dibromo-propylamine, hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8Br3N
CAS Number:
Molecular Weight:
297.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

BrCC(Br)CN.[H]Br

InChI

1S/C3H7Br2N.BrH/c4-1-3(5)2-6;/h3H,1-2,6H2;1H

InChI key

NQVQUJRBCVAMIL-UHFFFAOYSA-N

Application

In tandem with Turbo Grignard (Aldrich product 656984) and an electrophilic trap, 2,3-Dibromo-propylamine hydrobromide generates a "spring loaded" electrophile, which is a convenient reagent for the installation of azetidine to secondary amines. This product is one of several synthetic useful reagents reported by Baran and coworkers on Strain-release amination.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service