Skip to Content
Merck
All Photos(1)

Key Documents

A70607

Sigma-Aldrich

2-Amino-5-nitrophenol

technical grade, 90%

Synonym(s):

2-Hydroxy-4-nitroaniline, 3-Hydroxy-4-aminonitrobenzene, 3-Nitro-6-aminophenol, 5-Nitro-2-amino-1-hydroxybenzene, 5-Nitro-2-aminophenol, NSC 7087

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
972974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

90%

form

crystals

mp

198-202 °C (dec.) (lit.)

SMILES string

Nc1ccc(cc1O)[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-5-2-1-4(8(10)11)3-6(5)9/h1-3,9H,7H2

InChI key

DOPJTDJKZNWLRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Diazotation and coupling reactions
  • Preparation of biologically and pharmacologically active molecules

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Chen et al.
Cancer letters, 126(1), 67-74 (1998-05-01)
Two hair dye components, carcinogenic 4-nitro-2-aminophenol and 5-nitro-2-aminophenol, induced Cu(II)-dependent DNA cleavage frequently at thymine and guanine residues in DNA fragments obtained from the c-Ha-ras-1 protooncogene. When the p53 tumor suppressor gene was used, 4-nitro-2-aminophenol caused Cu(II)-dependent piperidine-labile sites at
2-Amino-5-nitrophenol.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 177-184 (1993-01-01)
Weixia Zhu et al.
Se pu = Chinese journal of chromatography, 30(9), 870-875 (2013-01-05)
An analytical method was developed for the simultaneous determination of 11 aminophenols in direct and oxidized hair dyes by high performance liquid chromatography (HPLC). The samples were extracted with methanol using sodium bisulfite for anti-oxidation. The purification was carried out
Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service