Skip to Content
Merck
All Photos(1)

Key Documents

94324

Sigma-Aldrich

Deoxo-Fluor solution

~50% in toluene (NMR)

Synonym(s):

Bis(2-methoxyethyl)aminosulfur trifluoride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14F3NO2S
CAS Number:
Molecular Weight:
221.24
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

concentration

~50% in toluene (NMR)

color

clear yellow

storage temp.

2-8°C

SMILES string

COCCN(CCOC)S(F)(F)F

InChI

1S/C6H14F3NO2S/c1-11-5-3-10(4-6-12-2)13(7,8)9/h3-6H2,1-2H3

InChI key

APOYTRAZFJURPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Fluorination reactions
  • Preparation of fluorinated catechols via Umpolung based nucleophilic deoxyfluorination
  • Preparation of imidazole derivatives as CXCR3 antagonists
  • Ring expansion reactions
  • Synthesis of acyl azides from carboxylic acids
  • Aldehydes or ketones from carboxylic acids and Grignard reagents via in situ generation of Weinreb amides

Other Notes

Fluorinating agent with high thermal stability [1][2][3][4]

Legal Information

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

<50.0 °F - closed cup

Flash Point(C)

< 10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajendra P Singh et al.
The Journal of organic chemistry, 68(15), 6063-6065 (2003-07-19)
New alpha,alpha-difluoroamides (4a-d, 6a-d) and alpha-ketoamides (5a-d, 7a-d) result from one-pot reactions of alpha-ketoacids, RCOCO(2)H (R = C(6)H(5), CH(3), CH(3)CH(2), thienyl) (1a-d) with bis(2-methoxyethyl)aminosulfur trifluoride [(CH(3)OCH(2)CH(2))(2)NSF(3)] (2) (Deoxofluor) or diethylaminosulfur trifluoride [(CH(3)CH(2))(2)NSF(3))] (3) (DAST). Product yields depend on reaction times
Fluorinating agent with high thermal stability.
S. Lal-Gauri et al.
The Journal of Organic Chemistry, 64, 7048-7048 (1999)
Lal et al.
The Journal of organic chemistry, 65(16), 4830-4832 (2000-08-24)
A variety of thiocarbonyl derivatives (thioketone, thioester, thioamide, dithioester, and dithiocarbamate) were converted to the corresponding gem-difluorides in excellent yields on reaction with the fluorinating agent, bis(2-methoxyethyl)aminosulfur trifluoride, in the presence of SbCl(3).
R.P. Singh, J.M. Shreeve
Journal of Fluorine Chemistry, 116, 23-23 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service