Skip to Content
Merck
All Photos(1)

Key Documents

92733

Sigma-Aldrich

Methyl 3-trimethylsiloxy-2-butenoate

technical, ~86% (trans + ~10% cis)

Synonym(s):

3-Trimethylsiloxy-2-butenoic acid methyl ester, Methyl 3-(trimethylsiloxy)crotonate, Methyl acetoacetate enol trimethylsilylether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O3Si
CAS Number:
Molecular Weight:
188.30
Beilstein:
2356944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

Assay

~86% (trans + ~10% cis)

refractive index

n20/D 1.447

bp

43-45 °C/0.3 mmHg (lit.)

density

0.966 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)\C=C(/C)O[Si](C)(C)C

Other Notes

Mild and efficient silylating agent[1]; Starting material to 1-methoxy-1,3-bis(trimethylsiloxy)butadiene; a diene for Diels-Alder reactions and an equivalent of the dianien of methyl acetoacetate[2][3][4]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Brownbridge et al.
Tetrahedron Letters, 4437-4437 (1979)
K. Yamamoto et al.
Chemistry Letters (Jpn), 649-649 (1978)
T.H. Chan et al.
Journal of the Chemical Society. Chemical Communications, 578-578 (1979)
T. Veysoglu et al.
Tetrahedron Letters, 22, 1303-1303 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service