Skip to Content
Merck
All Photos(2)

Documents

920479

Sigma-Aldrich

GPhos Pd G4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H68NO5PPdS
Molecular Weight:
920.53
UNSPSC Code:
12352103
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

GPhos Pd G4 is a fourth generation (G4) Buchwald palladium precatalyst for cross-coupling, demonstrating improved stability and reactivity from previous generations.

Learn more about G3 and G4 Buchwald Precatalysts

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott D McCann et al.
Journal of the American Chemical Society, 142(35), 15027-15037 (2020-08-14)
We have developed a new dialkylbiaryl monophosphine ligand, GPhos, that supports a palladium catalyst capable of promoting carbon-nitrogen cross-coupling reactions between a variety of primary amines and aryl halides; in many cases, these reactions can be carried out at room

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service