Skip to Content
Merck
All Photos(1)

Key Documents

919594

Sigma-Aldrich

Pd(IPr#)(AN)Cl2

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
£256.00
500 MG
£844.00

£256.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
£256.00
500 MG
£844.00

About This Item

Empirical Formula (Hill Notation):
C99H79Cl2N3Pd
Molecular Weight:
1488.03
UNSPSC Code:
12352200
NACRES:
NA.22

£256.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

Pd(IPr#)(AN)Cl2 is a Pd-catalyst that can be used in a broad range of reactions, including cross-coupling, C-H functionalization, electrophilic additions. NHC-Pd-aniline complexes of this type have many advantages, including higher reactivity in cross-coupling reactions than similar complexes using cinnamyl and pyridine throw-away ligands and simple reaction tunability through substituion of aniline ligand.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qun Zhao et al.
Chemical science, 12(31), 10583-10589 (2021-08-28)
IPr (IPr = 1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene) represents the most important NHC (NHC = N-heterocyclic carbene) ligand throughout the field of homogeneous catalysis. Herein, we report the synthesis, catalytic activity, and full structural and electronic characterization of novel, sterically-bulky, easily-accessible NHC ligands based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service