Skip to Content
Merck
All Photos(1)

Key Documents

901602

Sigma-Aldrich

Pivalic acid solution

1 M in dichloromethane

Synonym(s):

2,2-Dimethylpropionic acid, Trimethylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H10O2
CAS Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

description

fp: >135 deg °C

form

liquid

concentration

1 M in dichloromethane

refractive index

n/D 1.4204

density

1.260 g/mL

functional group

carboxylic acid

InChI

1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)

InChI key

IUGYQRQAERSCNH-UHFFFAOYSA-N

Application

Valuable building block offered as a solution in dichloromethane for more convenient handling.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service