Skip to Content
Merck
All Photos(2)

Key Documents

900654

Sigma-Aldrich

Poly(ε-caprolactone)-b-poly(ethylene glycol)-b-poly(ε-caprolactone)

5k-2k-5k

Synonym(s):

PCL-PEG-PCL, PCL-PEG-PCL triblock copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2CH2CH2CH2CO)mO(CH2CH2O)n(COCH2CH2CH2CH2CH2O)mH
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder or chunks

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Biocompatible, amphiphilic triblock polymer composed of a hydrophilic, central PEG block and two terminal hydrophobic PCL blocks. These materials have been used in drug delivery applications as well as thermoplastic elastomers. Well-defined materials with varying properties can be prepared by controlling the relative length of each polymer block. Hydroxyl termination allows for facile further chemical modification of these materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(caprolactone)-poly(ethylene glycol)-poly(caprolactone) (PCL-PEG-PCL) nanoparticles: a valuable and efficient system for in vitro and in vivo delivery of curcumin.
Manjii HK, et al.
Royal Society of Chemistry Advances, 6(17), 14403-14415 (2016)
ChangYang Gong et al.
International journal of pharmaceutics, 365(1-2), 89-99 (2008-09-17)
In this work, a series of biodegradable triblock poly(ethylene glycol)-poly(epsilon-caprolactone)-poly(ethylene glycol) (PEG-PCL-PEG, PECE) copolymers were successfully synthesized by ring-opening copolymerization, and were characterized by (1)H NMR, FT-IR, GPC, and DSC. Aqueous solutions of PECE copolymers underwent thermosensitive sol-gel-sol transition as
Synthesis and Characterization of PEG-PCL-PEG Triblock Copolymers as Carriers of Doxorubicin for the Treatment of Breast Cancer.
Cuong NV, et al.
Journal of Applied Polymer Science, 117, 3694-3703 (2010)
Synthesis and microphase separation of biodegradable poly(ε-caprolactone)-poly(ethylene glycol)-poly(ε-caprolactone) multiblock copolymer films.
You JH, et al.
Macromolecular Research, 16(7), 609-613 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service