Skip to Content
Merck
All Photos(1)

Key Documents

900166

Sigma-Aldrich

UV curable resin

greener alternative

adhesible to plastics and glass, low viscosity

Synonym(s):

CPS 1030 UV-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

viscosity

83 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)

density

1.11 g/mL

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35°C.
  • Keep tightly closed. Store in a cool, dry, well ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80°C), pressure and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and thus aligns with "Design for energy efficiency". Click here for more information.

Application

  • UV curable very low viscosity resin.
  • Cures rapidly to form a glassy high modulus material.
  • Excellent for use in applications requiring low viscosity, high hardness and low shrinkage stress.
  • Good for some spray coatings requiring no volatile organic compounds (VOCS).
  • Non yellowing.
  • Low oxygen inhibition.
  • Mechanical properties after curing:
Modulus*: 720MPa
Strength*: 28MPa
Elongation*: 7.4%
Tack free**: 10 fpm

*Test Resources: tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Which CAS# is addressed for UV RESIN? a bit confuses from the SDS as few CAS# listed down.

    1 answer
    1. The product itself is not assigned a CAS number. It is a combination of other chemicals, which have assigned CAS numbers, as outlined in section 3 of the SDS. This includes 42978-66-5, 40220-08-4, 7575-23-7, and 24615-84-7 with stabilizers 123-31-9, 150-76-5, and 98-29-3 also present. Please see the SDS, here: https://www.sigmaaldrich.com/sds/aldrich/900166

      Helpful?

  2. Please tell the refractive index of this 900166 product

    1 answer
    1. The refractive index of Product 900166 has not been measured, either for the liquid polymer as provided or for the resin formed after UV curing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service