Skip to Content
Merck
All Photos(1)

Key Documents

86876

Sigma-Aldrich

Tetrabutylammonium fluoride on Silica gel

extent of labeling: ~1.5 mmol/g F- loading

Synonym(s):

TBAF on silica gel, ‘Fluoride’ on silica gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

solid

extent of labeling

~1.5 mmol/g F- loading

Other Notes

Non hygroscopic, supported, ready-to-use source of "naked" fluoride ion; it is used in analogy to the unsupported reagent

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.H. Clark
Journal of the Chemical Society. Chemical Communications, 789-789 (1978)
A. Ricci et al.
Tetrahedron Letters, 23, 577-577 (1982)
A. Gambacorta et al.
Synthetic Communications, 19, 2441-2441 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service