Skip to Content
Merck
All Photos(1)

Key Documents

859869

Sigma-Aldrich

5-Thio-D-glucose

predominantly α-anomer, 96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5S
CAS Number:
Molecular Weight:
196.22
Beilstein:
1865193
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

96%

optical activity

[α]20/D +224°, c = 1 in 0.1 M HCl

mp

135-138 °C (lit.)

SMILES string

OC[C@H]1S[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O5S/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

KNWYARBAEIMVMZ-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service