Skip to Content
Merck
All Photos(1)

Key Documents

858803

Sigma-Aldrich

(2-Naphthoxy)acetic acid, N-hydroxysuccinimide ester

98%

Synonym(s):

N-Succinimidyl 2-naphthoxyacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO5
CAS Number:
Molecular Weight:
299.28
EC Number:
MDL number:
UNSPSC Code:
12352103

Quality Level

Assay

98%

mp

150-152 °C (lit.)

SMILES string

O=C(COc1ccc2ccccc2c1)ON3C(=O)CCC3=O

Application

Reagent for derivatization of amino acids and peptides.

Packaging

Packaged in glass bottles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service