Skip to Content
Merck
All Photos(1)

Documents

855316

Sigma-Aldrich

Nα-Acetyl-L-tyrosinamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CONH2
CAS Number:
Molecular Weight:
222.24
Beilstein:
2216288
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

optical activity

[α]21/D +51°, c = 0.8 in H2O

mp

223-225 °C (lit.)

SMILES string

CC(=O)N[C@@H](Cc1ccc(O)cc1)C(N)=O

InChI

1S/C11H14N2O3/c1-7(14)13-10(11(12)16)6-8-2-4-9(15)5-3-8/h2-5,10,15H,6H2,1H3,(H2,12,16)(H,13,14)/t10-/m0/s1

InChI key

RJNKBEQRBIJDNM-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service