Skip to Content
Merck
All Photos(1)

Documents

809225

Sigma-Aldrich

N-Ethyl-N-isopropylpropan-2-aminium 4-Oxo-3-(2,3,5,6-tetrafluoro-4-(methoxycarbonyl)phenyl)-1,5-dioxaspiro[5.5]undec-2-en-2-olate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H33F4NO6
CAS Number:
Molecular Weight:
519.53
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

117-122 °C

storage temp.

2-8°C

SMILES string

FC1=C(C(OC)=O)C(F)=C(F)C(C2=C([O-])OC3(CCCCC3)OC2=O)=C1F.CC[NH+](C(C)C)C(C)C

InChI

1S/C17H14F4O6.C8H19N/c1-25-14(22)9-12(20)10(18)7(11(19)13(9)21)8-15(23)26-17(27-16(8)24)5-3-2-4-6-17;1-6-9(7(2)3)8(4)5/h23H,2-6H2,1H3;7-8H,6H2,1-5H3

InChI key

CJYWUHPRPZRMSC-UHFFFAOYSA-N

General description

N-Ethyl-N-isopropylpropan-2-aminium 4-oxo-3-(2,3,5,6-tetrafluoro-4-(methoxycarbonyl)phenyl)-1,5-dioxaspiro[5.5]undec-2-en-2-olate is a fluoroarylated-Meldrum′s acid adduct. It can be prepared by reacting methyl-2,3,4,5,6-pentafluorobenzoate with cyclohexyl Meldrum′s acid.

Application

This compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameera M Senaweera et al.
The Journal of organic chemistry, 79(21), 10466-10476 (2014-10-02)
This work describes the facile and mono-selective per- and polyfluoroarylation of Meldrum's acid to generate a versatile synthon for highly fluorinated α-phenyl acetic acid derivatives, which provide straightforward access to fluorinated building blocks. The reaction takes place quickly, and most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service