Skip to Content
Merck
All Photos(1)

Key Documents

802190

Sigma-Aldrich

Poly(D,L-lactide-block-acrylic acid)

PDLLA average Mn 9,000, PAA average Mn 9,000

Synonym(s):

PLLA-block-PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H25S3(C3H4O2)xC6H9NO(C3H4O2)yH
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

PAA average Mn 9,000
PDLLA average Mn 9,000

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This diblock copolymer can be used to self-assemble into a drug delivery vehicle. This particular block ratio and molecular weight forms cylindrical micelles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service