Skip to Content
Merck
All Photos(1)

Documents

795801

Sigma-Aldrich

{[2-(Trifluoromethyl)phenyl]methyl}phosphonic acid

Synonym(s):

2-CF3BPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F3O3P
CAS Number:
Molecular Weight:
240.12
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

OP(CC1=C(C(F)(F)F)C=CC=C1)(O)=O

InChI

1S/C8H8F3O3P/c9-8(10,11)7-4-2-1-3-6(7)5-15(12,13)14/h1-4H,5H2,(H2,12,13,14)

InChI key

LJNXXFOSZJOZKN-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting and work function of metal oxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Hotchkiss et al.
Accounts of chemical research, 45(3), 337-346 (2011-10-21)
Transparent metal oxides, in particular, indium tin oxide (ITO), are critical transparent contact materials for applications in next-generation organic electronics, including organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs). Understanding and controlling the surface properties of ITO allows for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service