Skip to Content
Merck
All Photos(1)

Key Documents

776343

Sigma-Aldrich

DNTPD

99%

Synonym(s):

N4,N4-Bis[4-[bis(3-methylphenyl)amino]phenyl]-N4,N4-diphenyl-[1,1′-biphenyl]-4,4′-diamine, [1,1′-Biphenyl]-4,4′-diamine, N,N′-bis[4-[bis(3-methylphenyl)amino]phenyl]-N,N′-diphenyl-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H54N4
CAS Number:
Molecular Weight:
879.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

105-112 °C

λmax

326 nm in THF

SMILES string

Cc1cccc(c1)N(c2ccc(cc2)N(c3ccccc3)c4ccc(cc4)-c5ccc(cc5)N(c6ccccc6)c7ccc(cc7)N(c8cccc(C)c8)c9cccc(C)c9)c%10cccc(C)c%10

InChI

1S/C64H54N4/c1-47-15-11-23-61(43-47)67(62-24-12-16-48(2)44-62)59-39-35-57(36-40-59)65(53-19-7-5-8-20-53)55-31-27-51(28-32-55)52-29-33-56(34-30-52)66(54-21-9-6-10-22-54)58-37-41-60(42-38-58)68(63-25-13-17-49(3)45-63)64-26-14-18-50(4)46-64/h5-46H,1-4H3

InChI key

SPDPTFAJSFKAMT-UHFFFAOYSA-N

General description

DNTPD is a hole transporting material with good optical properties. It has high ionization energy (IE) and hole mobility which allow excellent performance for electron blocking.[1][2]

Application

DNTPD can be coated on indium tin oxide (ITO) to form an optimized device with an electron transporting layer for the fabrication of high efficiency organic light emitting diodes (OLEDs).[3][4][5]
Hole Injection/Transport Materials for OLEDs

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High hole mobility hole transport material for organic light-emitting devices
Huh DH, et al.
Synthetic Metals, 180, 79-84 (2013)
Synt. Metals, 161, 2024-2030 (2011)
Ideal host and guest system in phosphorescent OLEDs
Jeon WS, et al.
Organic Electronics, 10(2), 240-246 (2009)
Molecular Engineering of Blue Fluorescent Molecules Based on Silicon End-Capped Diphenylaminofluorene Derivatives for Efficient Organic Light-Emitting Materials
Lee KH, et al.
Advances in Functional Materials, 20(8), 1345-1358 (2010)
A Pyridine-Containing Anthracene Derivative with High Electron and Hole Mobilities for Highly Efficient and Stable Fluorescent Organic Light-Emitting Diodes
Sun Y, et al.
Advances in Functional Materials, 21(10), 1881-1886 (2011)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service