Skip to Content
Merck
All Photos(2)

Documents

768723

Sigma-Aldrich

3-Butenyl 2-(dodecylthiocarbonothioylthio)-2-methylpropionate

97%

Synonym(s):

-ene functional CTA, alkene RAFT agent, butene RAFT agent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38O2S3
Molecular Weight:
418.72
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.526

density

1.011 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=CCCOC(C(C)(C)SC(SCCCCCCCCCCCC)=S)=O

InChI

1S/C21H38O2S3/c1-5-7-9-10-11-12-13-14-15-16-18-25-20(24)26-21(3,4)19(22)23-17-8-6-2/h6H,2,5,7-18H2,1,3-4H3

InChI key

PJYDHYBGYFGBRC-UHFFFAOYSA-N

General description

Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Butene can be used to react with thiols for thiol-ene ligation or cyclopentadiene/furan to form norbornene structures for eventual ROMP reactions. This can also be used Chain Transfer Agent (CTA)
Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service