Skip to Content
Merck
All Photos(1)

Key Documents

758132

Sigma-Aldrich

6-Amino-2,3-dimethylquinoxaline

95%

Synonym(s):

2,3-Dimethylquinoxalin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

Assay

95%

form

solid

mp

190-195 °C

SMILES string

Cc1nc2ccc(N)cc2nc1C

InChI

1S/C10H11N3/c1-6-7(2)13-10-5-8(11)3-4-9(10)12-6/h3-5H,11H2,1-2H3

InChI key

QBZGAULXCVZXFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service