Skip to Content
Merck
All Photos(1)

Key Documents

754722

Sigma-Aldrich

Thiophene-2-boronic acid pinacol ester

98% (GC)

Synonym(s):

4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15BO2S
CAS Number:
Molecular Weight:
210.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (GC)

form

powder or crystals

mp

65-70 °C

λmax

239 nm at 0.01 g/L in methanol

SMILES string

CC1(C)OB(OC1(C)C)c2cccs2

InChI

1S/C10H15BO2S/c1-9(2)10(3,4)13-11(12-9)8-6-5-7-14-8/h5-7H,1-4H3

InChI key

FFZHICFAHSDFKZ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim, J.; et al
Journal of Polymer Science Part A: Polymer Chemistry, 49, 369-380 (2011)
Park, J.-H; et al.
Organic Electrochemistry, 11, 820-830 (2010)
John Jun-An Chen et al.
ACS applied materials & interfaces, 2(9), 2679-2686 (2010-09-02)
New soluble quinacridone-based molecules have been developed as electron donor materials for solution-processed organic solar cells. By functionalizing the pristine pigment core of quinacridone with solubilizing alkyl chains and light absorbing/charge transporting thiophene units, i.e., bithiophene (BT) and thienylbenzo[c][1,2,5]thiadiazolethienyl (BTD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service